Assay Details
| Gene Symbol: TERT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046242.1 | NP_001039707.1 | 10 - 11 | 2640 | 62 | |
| GenBank mRNA | DQ464898.1 | - | 10 - 11 | 2640 | 62 | |
| DQ464899.1 | - | 10 - 11 | 2640 | 62 | ||
More Information
Set Membership: |
|

TERT
Genomic Map