Assay Details
| Gene Symbol: VPS35 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046258.1 | NP_001039723.1 | 2 - 3 | 133 | 68 | |
| GenBank mRNA | BC105430.1 | - | 2 - 3 | 133 | 68 | |
More Information
Set Membership: |
|

VPS35
Genomic Map