Assay Details
| Gene Symbol: DPM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046268.2 | NP_001039733.1 | 2 - 3 | 108 | 82 | |
| GenBank mRNA | BC112577.1 | - | 2 - 3 | 109 | 82 | |
More Information
Set Membership: |
|

DPM2
Genomic Map