Assay Details
| Gene Symbol: CLN5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046299.1 | NP_001039764.1 | 1 - 2 | 169 | 98 | |
| GenBank mRNA | DQ421787.1 | - | 1 - 2 | 169 | 98 | |
More Information
Set Membership: |
|

CLN5
Genomic Map