Assay Details
| Gene Symbol: ELAC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046327.1 | NP_001039792.1 | 2 - 3 | 628 | 107 | |
| GenBank mRNA | BC113356.1 | - | 2 - 3 | 628 | 107 | |
More Information
Set Membership: |
|

ELAC1
Genomic Map