Assay Details
| Gene Symbol: BPNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046335.1 | NP_001039800.1 | 1 - 2 | 633 | 84 | |
| GenBank mRNA | BC105514.1 | - | 1 - 2 | 633 | 84 | |
More Information
Set Membership: |
|

BPNT2
Genomic Map