Assay Details
| Gene Symbol: BPNT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046335.1 | NP_001039800.1 | 2 - 3 | 793 | 80 | |
| GenBank mRNA | BC105514.1 | - | 2 - 3 | 793 | 80 | |
More Information
Set Membership: |
|

BPNT2
Genomic Map