Assay Details
| Gene Symbol: RITA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046344.1 | NP_001039809.1 | 1 - 2 | 228 | 64 | |
| GenBank mRNA | BC113269.1 | - | 1 - 2 | 228 | 64 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC113269.1, NM_001046344.1 transcripts.
Set Membership: |
|

RITA1
Genomic Map