Assay Details
| Gene Symbol: UGT2B10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046354.2 | NP_001039819.1 | 2 - 3 | 900 | 91 | |
| GenBank mRNA | BC112641.1 | - | 2 - 3 | 901 | 91 | |
More Information
Set Membership: |
|

UGT2B10
Genomic Map