Assay Details
| Gene Symbol: CMTM8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046358.1 | NP_001039823.1 | 1 - 2 | 243 | 86 | |
| GenBank mRNA | BC114787.1 | - | 1 - 2 | 243 | 86 | |
More Information
Set Membership: |
|

CMTM8
Genomic Map