Assay Details
| Gene Symbol: LYPD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046359.1 | NP_001039824.1 | 2 - 3 | 467 | 79 | |
| GenBank mRNA | BC114130.1 | - | 2 - 3 | 467 | 79 | |
More Information
Set Membership: |
|

LYPD1
Genomic Map