Assay Details
| Gene Symbol: LRRC42 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046386.1 | NP_001039851.1 | 2 - 3 | 565 | 73 | |
| GenBank mRNA | BC113249.1 | - | 2 - 3 | 565 | 73 | |
More Information
Set Membership: |
|

LRRC42
Genomic Map