Assay Details
| Gene Symbol: CYLD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046417.1 | NP_001039882.1 | 3 - 4 | 982 | 71 | |
| GenBank mRNA | BC114710.1 | - | 3 - 4 | 982 | 71 | |
More Information
Set Membership: |
|

CYLD
Genomic Map