Assay Details
| Gene Symbol: G3BP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046455.1 | NP_001039920.1 | 7 - 8 | 940 | 78 | |
| GenBank mRNA | BC105567.1 | - | 7 - 8 | 940 | 78 | |
More Information
Set Membership: |
|

G3BP2
Genomic Map