Assay Details
| Gene Symbol: NSRP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046462.1 | NP_001039927.1 | 3 - 4 | 202 | 62 | |
| GenBank mRNA | BC112694.1 | - | 3 - 4 | 202 | 62 | |
More Information
Set Membership: |
|

NSRP1
Genomic Map