Assay Details
| Gene Symbol: GMEB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046475.1 | NP_001039940.1 | 5 - 6 | 672 | 71 | |
| GenBank mRNA | BC113252.1 | - | 5 - 6 | 672 | 71 | |
More Information
Set Membership: |
|

GMEB1
Genomic Map