Assay Details
| Gene Symbol: VAMP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046476.1 | NP_001039941.1 | 2 - 3 | 177 | 89 | |
| GenBank mRNA | BC112840.1 | - | 2 - 3 | 177 | 89 | |
More Information
Set Membership: |
|

VAMP5
Genomic Map