Assay Details
| Gene Symbol: MSN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046477.1 | NP_001039942.1 | 2 - 3 | 260 | 62 | |
| GenBank mRNA | BC113313.1 | - | 2 - 3 | 260 | 62 | |
More Information
Set Membership: |
|

MSN
Genomic Map