Assay Details
| Gene Symbol: EAF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046507.2 | NP_001039972.1 | 1 - 2 | 150 | 61 | |
| GenBank mRNA | BC112586.1 | - | 1 - 2 | 151 | 61 | |
More Information
Set Membership: |
|

EAF2
Genomic Map