Assay Details
| Gene Symbol: TSPAN12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046512.2 | NP_001039977.1 | 1 - 2 | 131 | 55 | |
| GenBank mRNA | BC114167.1 | - | 1 - 2 | 132 | 55 | |
More Information
Set Membership: |
|

TSPAN12
Genomic Map