Assay Details
| Gene Symbol: MAT2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046526.2 | NP_001039991.1 | 5 - 6 | 768 | 76 | |
| GenBank mRNA | BC114151.1 | - | 5 - 6 | 769 | 76 | |
More Information
Set Membership: |
|

MAT2B
Genomic Map