Assay Details
| Gene Symbol: TRMT13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046530.2 | NP_001039995.2 | 6 - 7 | 623 | 105 | |
| GenBank mRNA | BC114134.1 | - | 6 - 7 | 105 | ||
More Information
Set Membership: |
|

TRMT13
Genomic Map