Assay Details
| Gene Symbol: SNX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046533.2 | NP_001039998.1 | 3 - 4 | 406 | 51 | |
| GenBank mRNA | BC112867.1 | - | 3 - 4 | 407 | 51 | |
More Information
Set Membership: |
|

SNX8
Genomic Map