Assay Details
| Gene Symbol: TUBB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046549.2 | NP_001040014.1 | 2 - 3 | 340 | 75 | |
| GenBank mRNA | BC105401.1 | - | 2 - 3 | 341 | 75 | |
More Information
Set Membership: |
|

TUBB
Genomic Map