Assay Details
| Gene Symbol: LSM5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046553.2 | NP_001040018.1 | 4 - 5 | 262 | 132 | |
| GenBank mRNA | BC105404.1 | - | 4 - 5 | 263 | 132 | |
More Information
Set Membership: |
|

LSM5
Genomic Map