Assay Details
| Gene Symbol: RBMXL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046610.1 | NP_001040075.1 | 1 | 1453 | 92 | |
| GenBank mRNA | BC114040.1 | - | 1 | 1453 | 92 | |
More Information
Set Membership: |
|

RBMXL2
Genomic Map