Assay Details
| Gene Symbol: ZNF3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046614.1 | NP_001040079.1 | 5 - 6 | 589 | 89 | |
| GenBank mRNA | BC105476.1 | - | 5 - 6 | 589 | 89 | |
More Information
Set Membership: |
|

ZNF3
Genomic Map