Assay Details
| Gene Symbol: TMEM50A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075163.1 | NP_001068631.1 | 4 - 5 | 364 | 77 | |
| GenBank mRNA | BC118224.1 | - | 4 - 5 | 364 | 77 | |
| BC151415.1 | - | 4 - 5 | 385 | 77 | ||
More Information
Set Membership: |
|

TMEM50A
Genomic Map