Assay Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075234.1 | NP_001068702.1 | 2 - 3 | 230 | 75 | |
| GenBank mRNA | BC116019.1 | - | 2 - 3 | 230 | 75 | |
| BT029897.1 | - | 1 - 2 | 168 | 75 | ||
More Information
Set Membership: |
|

MYL9
Genomic Map