Assay Details
| Gene Symbol: WBP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075251.1 | NP_001068719.1 | 2 - 3 | 223 | 81 | |
| GenBank mRNA | BC118233.1 | - | 2 - 3 | 223 | 81 | |
More Information
Set Membership: |
|

WBP4
Genomic Map