Assay Details
| Gene Symbol: TFPT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075274.2 | NP_001068742.1 | 4 - 5 | 488 | 82 | |
| GenBank mRNA | AM930552.2 | - | 2 - 3 | 106 | 82 | |
| BC118355.1 | - | 4 - 5 | 489 | 82 | ||
More Information
Set Membership: |
|

TFPT
Genomic Map