Assay Details
| Gene Symbol: NUP54 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075285.1 | NP_001068753.1 | 11 - 12 | 1454 | 63 | |
| GenBank mRNA | BC114015.1 | - | 11 - 12 | 1454 | 63 | |
More Information
Set Membership: |
|

NUP54
Genomic Map