Assay Details
| Gene Symbol: AP3S2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075376.1 | NP_001068844.1 | 1 - 2 | 110 | 59 | |
| GenBank mRNA | BC116115.1 | - | 1 - 2 | 110 | 59 | |
| BT025371.1 | - | 1 - 2 | 86 | 59 | ||
More Information
Set Membership: |
|

AP3S2
Genomic Map