Assay Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075391.2 | NP_001068859.1 | 1 - 2 | 163 | 55 | |
| GenBank mRNA | BC114197.1 | - | 1 - 2 | 164 | 55 | |
More Information
Set Membership: |
|

SPC24
Genomic Map