Assay Details
| Gene Symbol: SPC24 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075391.2 | NP_001068859.1 | 3 - 4 | 416 | 83 | |
| GenBank mRNA | BC114197.1 | - | 3 - 4 | 417 | 83 | |
More Information
Set Membership: |
|

SPC24
Genomic Map