Assay Details
| Gene Symbol: SLC25A39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075415.1 | NP_001068883.1 | 5 - 6 | 421 | 62 | |
| GenBank mRNA | BC118337.1 | - | 5 - 6 | 421 | 62 | |
More Information
Set Membership: |
|

SLC25A39
Genomic Map