Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075426.1 | NP_001068894.1 | 4 - 5 | 1599 | 84 | |
| GenBank mRNA | BC114763.1 | - | 4 - 5 | 1599 | 84 | |
| EU599224.1 | - | 4 - 5 | 1331 | 84 | ||
More Information
Set Membership: |
|

ANGPTL1
Genomic Map