Assay Details
| Gene Symbol: CIB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075433.2 | NP_001068901.1 | 3 - 4 | 312 | 59 | |
| GenBank mRNA | BC118410.1 | - | 3 - 4 | 313 | 59 | |
More Information
Set Membership: |
|

CIB1
Genomic Map