Assay Details
| Gene Symbol: MYBL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075448.2 | NP_001068916.1 | 12 - 13 | 2058 | 57 | |
| GenBank mRNA | BC118123.1 | - | 12 - 13 | 2059 | 57 | |
More Information
Set Membership: |
|

MYBL2
Genomic Map