Assay Details
| Gene Symbol: DBX2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075461.1 | NP_001068929.1 | 3 - 4 | 1010 | 70 | |
| GenBank mRNA | BC118220.1 | - | 3 - 4 | 1010 | 70 | |
More Information
Set Membership: |
|

DBX2
Genomic Map