Assay Details
| Gene Symbol: PON3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075479.1 | NP_001068947.1 | 3 - 4 | 212 | 70 | |
| GenBank mRNA | BC116035.1 | - | 3 - 4 | 212 | 70 | |
More Information
Set Membership: |
|

PON3
Genomic Map