Assay Details
| Gene Symbol: AGA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075511.2 | NP_001068979.1 | 9 - 10 | 996 | 118 | |
| GenBank mRNA | BC114812.1 | - | 9 - 10 | 997 | 118 | |
More Information
Set Membership: |
|

AGA
Genomic Map