Assay Details
| Gene Symbol: GNS | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075562.2 | NP_001069030.1 | 11 - 12 | 1408 | 64 | |
| GenBank mRNA | BC115990.1 | - | 11 - 12 | 1409 | 64 | |
More Information
Set Membership: |
|

GNS
Genomic Map