Assay Details
| Gene Symbol: KCNIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075612.2 | NP_001069080.1 | 5 - 6 | 517 | 65 | |
| GenBank mRNA | BC118417.1 | - | 5 - 6 | 518 | 65 | |
More Information
Set Membership: |
|

KCNIP3
Genomic Map