Assay Details
| Gene Symbol: PAGR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075632.1 | NP_001069100.1 | 2 - 3 | 1041 | 77 | |
| GenBank mRNA | BC118095.1 | - | 2 - 3 | 1041 | 77 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118095.1, NM_001075632.1 transcripts.
Set Membership: |
|

PAGR1
Genomic Map