Assay Details
| Gene Symbol: MST1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075677.2 | NP_001069145.1 | 1 - 2 | 81 | 70 | |
| GenBank mRNA | BC114002.1 | - | 1 - 2 | 82 | 70 | |
More Information
Set Membership: |
|

MST1
Genomic Map