Assay Details
| Gene Symbol: GPRC5B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075741.1 | NP_001069209.1 | 2 - 3 | 1119 | 77 | |
| GenBank mRNA | BC149136.1 | - | 2 - 3 | 1103 | 77 | |
| BT025414.1 | - | 2 - 3 | 1119 | 77 | ||
More Information
Set Membership: |
|

GPRC5B
Genomic Map