Assay Details
| Gene Symbol: TBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075742.1 | NP_001069210.1 | 4 - 5 | 819 | 87 | |
| GenBank mRNA | BC113308.1 | - | 4 - 5 | 819 | 87 | |
More Information
Set Membership: |
|

TBP
Genomic Map