Assay Details
| Gene Symbol: CFAP410 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075781.1 | NP_001069249.1 | 3 - 4 | 215 | 93 | |
| GenBank mRNA | BC118255.1 | - | 3 - 4 | 215 | 93 | |
More Information
Set Membership: |
|

CFAP410
Genomic Map