Assay Details
| Gene Symbol: DNASE2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075865.1 | NP_001069333.1 | 1 - 2 | 153 | 84 | |
| GenBank mRNA | BC118435.1 | - | 1 - 2 | 153 | 84 | |
More Information
Set Membership: |
|

DNASE2B
Genomic Map